You searched for: Safety Cans
Holding corrosives or flammable liquids such as paint thinner and lighter fluid, the leak-proof safety cans are designed to comply with safety codes and regulations. Easy-to-fill designs with tight seal closures provide maximum protection for the environment and employees. Able to withstand physical shock encountered during transportation, safety cans store and dispose solvents safely. Select models feature a hose for direct pouring or carbon filters that trap chemical vapors from being inhaled.
VWR® Solvent Waste System
Supplier: VWR International
This solvent waste system is designed to be an affordable solution for HPLC liquid waste disposal.
Expand 4 Items
Paint Cans, Epoxy Lined, Triple Tite Lid, Metal, Qorpak®
Supplier: Qorpak
Epoxy Lined Round Paint Can with Triple Tite Lid provide a non-reactive barrier between the container and the product/atmosphere; protecting the contents and preventing rust
Expand 3 Items
Aerosol Can Recycler, PIG
Supplier: New Pig
Recycling system easily punctures aerosol cans, rendering them exempt from hazardous waste regulations when properly emptied.
Expand 1 Items
Waste Systems, Canister Kits, S.C.A.T.
Supplier: S.C.A.T.
Complete waste systems including canister, SafetyWasteCap and exhaust filter.
Expand 3 Items
SP Bel-Art Safety Waste Jug, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
For safe, in-plant handling and temporary storage of spent solvents or other liquid wastes awaiting proper disposal.
Expand 1 Items
F-Style Cans, Metal
Supplier: Qorpak
Metal F-Style Oblong cans are ideal for products such as cooking oils, paint thinners, turpentine, lighter fluids, insecticides, and general storage applications.
Expand 5 Items
Liquid Disposal Safety Cans, Justrite®
Supplier: Justrite
These high-density polyethylene (HDPE) disposal cans with built-in fill gauge provide superior chemical resistance and leak protection.
Expand 2 Items
Safety Dispensing Cans, Justrite®
Supplier: Justrite
Steel laboratory safety cans offer reliable dispensing features at an economical price.
Expand 14 Items
Safety Containers, Justrite®
Supplier: Justrite
Safely contain hazardous liquids and control vapors to reduce the risk of fire, protecting both personnel and facilities.
Expand 11 Items
Safety Cans with Faucet, Eagle Manufacturing
Supplier: Eagle Manufacturing
For controlled dispensing of flammable liquids into small vessels using built-in, self-closing faucet valve.
Expand 2 Items
Laboratory Safety Cans, Eagle Manufacturing
Supplier: Eagle Manufacturing
For safe transport and dispensing of flammable liquids.
Expand 1 Items
Safety Plunger Cans, Eagle Manufacturing
Supplier: Eagle Manufacturing
For safe cleaning with flammable or volatile liquids.
Expand 6 Items
Type I Nonmetallic Safety Cans, Justrite®
Supplier: Justrite
High-density polyethylene cans are dent-, puncture-, and chip-resistant
Expand 4 Items
Type II AccuFlow™ Safety Cans for various types of liquids, Justrite®
Supplier: Justrite
Going beyond protection and compliance, AccuFlow™ Type II Safety Cans provide fast, precision pouring when handling flammable liquids
Expand 18 Items
Type I Safety Cans for various types of liquids, Justrite®
Supplier: Justrite
Compliant and economical Justrite™ Type I safety cans reduce fire risks and make everyday use of flammables easier and safer
Expand 24 Items
Centura® Prefabricated Quick-Disconnect Safety Disposal Cans, Justrite®
Supplier: Justrite
Ideal for HPLC applications, these durable polyethylene safety cans offer a hands-free way to safely collect liquid waste.
Expand 7 Items
Nonmetallic Laboratory Safety Cans, Justrite®
Supplier: Justrite
High-density polyethylene safety cans for use on shelves or in tilt stands have self-closing faucet for convenient, controlled dispensing of hazardous liquids. Fill spout has self-closing pressure relief cap and double mesh flame arrester. The 18.9L (5gal.) cans have built-in vacuum breakers. Oval-shaped safety can 56609-586 has stainless steel faucet and round can 56609-589 has brass faucet, both mounted 5cm (2") above bottom. Tilt can 56609-588 with top faucet rests on a sturdy metal cradle which measures 36.8x35.5cm (14½x14") at the base.
Expand 1 Items
Safety Disposal Cans, Type 316 Stainless Steel, Eagle Manufacturing
Supplier: Eagle Manufacturing
Designed for use as an intermediate station for liquid wastes.
Expand 2 Items
Type I Safety Cans, Justrite®
Supplier: Justrite
Designed for carrying flammable liquids to pour into containers with wider openings.
Expand 1 Items
Type I Safety Cans, Galvanized Steel, Eagle Manufacturing
Supplier: Eagle Manufacturing
For carrying and dispensing flammable liquids safely.
Expand 9 Items
Type II Safety Cans, Galvanized Steel, Eagle Manufacturing
Supplier: Eagle Manufacturing
For safe transport, storage, and dispensing of flammable liquids.
Expand 2 Items
Type I Safety Cans for Flammables, Justrite®
Supplier: Justrite
Compliant and economical Justrite™ Type I safety cans reduce fire risks and make everyday use of flammables easier and safer.
Expand 7 Items
Bench and Daub Safety Can, Eagle Manufacturing
Supplier: Eagle Manufacturing
For safer cleaning of small parts or swabbing larger ones
Expand 5 Items
HPLC Safety Disposal Cans, Justrite®
Supplier: Justrite
Keep laboratory and personnel safe with a compliant, environmentally-friendly solution to waste collection.
Expand 2 Items
Type I Poly Safety Cans, Justrite®
Supplier: Justrite
Chemical-resistance HDPE cans offer safe storage for flammable corrosives.
Expand 1 Items
Plunger Cans, Justrite®
Supplier: Justrite
Plunger can helps moisten cleaning cloths or swabs quickly and easily
Expand 5 Items
Bench Cans, Justrite®
Supplier: Justrite
Terne plate steel bench can for convenient cleaning of small parts in solvents
Expand 6 Items
Nonmetallic Dispenser Cans, Justrite®
Supplier: Justrite
For easy one-hand dispensing of flammable liquids directly onto work
Expand 3 Items
Safety Disposal Cans, Galvanized Steel, Eagle Manufacturing
Supplier: Eagle Manufacturing
These galvanized steel disposable cans have a red powder coat finish
Expand 2 Items
Justrite® Liquid Disposal Can and ECO Funnels
Supplier: Justrite
A safe solution for containing flammable wastes and corrosives, this Justrite Liquid Disposal Can is constructed of high density polyethylene for superior chemical resistance and leak protection.