You searched for: Membrane Filters
Membrane filters serve as effective physical barriers that remove solids, viruses, bacteria, and other unwanted molecules. Specialized types are designed for softening, disinfecting, organic removal processing, and water desalination. The filters can be installed in compact, automated, or modular units. These strainers can be used as separators between stacked discs in a single filter holder during serial filtration. Membrane filters are virtually inert to chemical and biological agents, ensuring stability when filtering concentrated acids, alkalis, or oxidizers.
wwPTFE (Water Wettable PTFE) Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Next generation of universal hydrophilic PTFE membrane with maximum chemical compatibility when filtering both aqueous solutions and aggressive solvents.
Expand 7 Items
Nylaflo® Membrane Disc Filters
Supplier: Cytiva
Useful for a wide range of both aqueous and solvent-based applications.
Expand 9 Items
Metricel® Black Membrane Disc Filters
Supplier: Cytiva
Ideal for the isolation and enumeration of yeast and mold colonies from water and beverages using the Membrane Filter Technique referenced in Standard Methods for the Examination of Water and Wastewater.
Expand 4 Items
Metricel® Polypropylene Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Pure polypropylene membranes for filtration of aggressive solvents.
Expand 1 Items
GN Metricel® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The GN-6 membrane is certified for the microbiological analysis of potable, waste, process, and natural waters in accordance with the Membrane Filter Technique referenced in Standard Methods for the Examination of Water and Wastewater.
Expand 10 Items
Membrane Disc Filters, GLA-5000, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Ideal for sampling airborne metals, silica, and dust.
Expand 4 Items
Ultrafiltration Membrane Disc Filters
Supplier: Cytiva
Replacement membranes for conventional, nondisposable stirred cell systems.
Expand 24 Items
Separator Depth Filters, Polypropylene/Polyethylene, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The Polypropylene/Polyethylene Separator Depth Filters have outstanding wet strength and can be folded or handled the same as laboratory paper. Because of their material of construction, these disc filters are virtually inert to chemical and biological agents.
Expand 2 Items
Versapor® Membrane Disc Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Ideal for prefiltration of difficult-to-filter solutions and serum.
Expand 15 Items
Teflo Air Sampling Membranes, PTFE, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Strong, chemically resistant PTFE membranes have unique PMP (polymethylpentene) support ring for PM10 and PM2.5 dichotomous and other air sampling techniques.
Expand 2 Items
Dispenser Refill Packs for Sentino® Filter Dispenser, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These membranes have been designed specifically for use with the Sentino® automatic filter dispenser.
Expand 4 Items
Supor® Membrane Disc Filters
Supplier: Cytiva
Supor® hydrophilic polyethersulfone membranes offer higher fluid flow rates, lower protein binding, lower extractables, and greater strength than other cellulosic (acetate, nitrate) membranes.
Expand 35 Items
Ultipor® Nylon Membrane Filter Discs, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Ultipor® Nylon Filter Discs are made of a reinforced single layer of N66 Nylon with nonwoven internal polyester.
Expand 1 Items
Nalgene® Filter Membranes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Filter membranes are ideal for use in particulate and microbial analysis.
Expand 4 Items
Nalgene® Water Quality Membrane, Cellulose Nitrate, Gridded, Thermo Scientific
Supplier: Thermo Fisher Scientific
Membrane turns black when wet.
Expand 2 Items
Membrane Filters with Grid, Sartorius
Supplier: Sartorius
Hydrophobic cellulose nitrate and cellulose acetate gridded membrane filters for colony count and sterility testing, with outstanding retention rates for microorganisms.
Expand 14 Items
Membrane Filters, Type 113, Sartorius
Supplier: Sartorius
Cellulose nitrate hydrophilic membrane filters. For sample pretreatment, particle testing and chemotaxis.
Expand 10 Items
Membrane Filters with Grid, Sartorius
Supplier: Sartorius
Cellulose nitrate hydrophilic, gridded membrane filters for colony counting, particle testing and microscopy, with outstanding recovery rates for microorganisms.
Expand 39 Items
Microsart® e.motion Membranes, Sartorius
Supplier: Sartorius
These sterile cellulose nitrate and PES Microsart® e.motion membrane filters can be used for colony counting, particle testing and microscopy. They are specifically designed for use in the Microsart® e.motion dispenser.
Expand 17 Items
Whatman™ Cyclopore PC Membranes
Supplier: Cytiva
Cytiva's Whatman Cyclopore polycarbonate membranes are true pore size microporous membranes featuring the sharp cut-off and reproducible microfiltration characteristics of track-etched membranes.
Expand 8 Items
Whatman™ Nuclepore Hydrophilic Membrane, Whatman products (Cytiva)
Supplier: Cytiva
Nuclepore™ track-etched hydrophilic membrane filters offer a highly defined pore size cut-off in the submicron range, enabling particle deformity measurement and capture of extremely small particles. The membranes have sharply defined pores, high flow rates, and excellent chemical and thermal resistance. They are suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
Expand 23 Items
Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membranes
Supplier: Cytiva
Nuclepore™ track-etched hydrophilic membrane filters offer a highly defined pore size cut-off in the submicron range, enabling particle deformity measurement and capture of extremely small particles. The membranes have sharply defined pores, high flow rates, and excellent chemical and thermal resistance. They are suitable for a variety of samples and applications, including microscopy, cytology, parasitology, and certain air and oceanography analyses.
Expand 38 Items
Cellulose Acetate Membrane, Apacor
Supplier: Apacor
Membranes are designed for use in the electrophoretic separations of serum proteins.
Expand 4 Items
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 0.45 µm
Supplier: Cytiva
Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
Expand 25 Items
Whatman™ Cellulose Acetate Membrane Filters, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman cellulose acetate membrane filters have very low protein binding, which minimizes sample loss when filtering protein-based aqueous samples.
Expand 16 Items
Whatman™ Regenerated Cellulose Membranes, Circles, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman regenerated cellulose membrane filters are microporous cellulose acetate filters made without wetting agents. RC60 offers low extractables and wide compatibility in both organic and aqueous media.
Expand 11 Items
Whatman™ PM2.5 Air Monitoring Membrane Filters, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman PM2.5 PTFE membrane filters are designed for PM 2.5 ambient air monitoring. These high-purity, thin filters are sequentially numbered and come with chemically resistant polypropylene support rings.
Expand 1 Items
Whatman™ Mixed Cellulose Ester Membranes, Sterile, 1.2 µm, Whatman products (Cytiva)
Supplier: Cytiva
Mixed cellulose ester, WME range, circles, gridded. Whatman mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
Expand 1 Items
Whatman™ MembraClear Cellulose Ester Membrane Filters, Whatman products (Cytiva)
Supplier: Cytiva
These mixed cellulose ester membrane filters are designed for asbestos sampling using the membrane filter method for phase contrast microscopy. Asbestos sampling isolates these fibers from circulating air to determine concentrations.