ContactUsLinkComponent
125 results for Retort Stands
You searched for: Retort Stands
Retort Stands
Retort stands to hold burettes, flasks, and other labware during procedures are available at VWR. Sets include both stainless steel rod and base, constructed of your choice of plastic or cast iron. A polypropylene retort stand is offered for severe corrosive conditions. Bases of cast iron coated with enamel are featured in rectangular, tripod, U-shaped and A-shaped options. Different rod lengths for the support of different diameters of labware are also supplied through VWR, as well as retort rings and clamps.
Sort by
VWR® Talon® Gas Cylinder Stand
VWR® Talon® Gas Cylinder Stand
Supplier: VWR International
Permits convenient, safe cylinder transfer and upright support.
Expand 1 Items
Loading...
Funnel Support
Funnel Support
Supplier: VWR International
With iron clamp for support rods of any size to 5/8". The four holes are beveled and slotted and are spaced at 41/2" center distance.
Expand 1 Items
Loading...
VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
Supplier: VWR International
Support stand base is constructed of cast iron with black enamel finish.
Expand 1 Items
Loading...
VWR® Talon® Support
VWR® Talon® Support
Supplier: VWR International
Multi-purpose support was originally designed to permit cylindrical vessels up to 305mm (12") in diameter to sit close to the upright rod.
Expand 1 Items
Loading...
VWR® Talon® Support Plates
VWR® Talon® Support Plates
Supplier: VWR International
Designed to hold hotplate/stirrers and other apparatus to lab frames or ring stands.
Expand 3 Items
Loading...
Rectangular Bases
Supplier: VWR International
Essential for Any Chemistry or Physics Laboratory
Expand 3 Items
Loading...
Rod and Base Assembly
Rod and Base Assembly
Supplier: VWR International
This assembly is handy for multiple lab applications.
Expand 3 Items
Loading...
A-Frame Base
A-Frame Base
Supplier: VWR International
This cast iron base features threads for rods sized 1/2 - 13.
Expand 2 Items
Loading...
VWR® Talon® Support Stands
VWR® Talon® Support Stands
Supplier: VWR International
Support stands are made entirely of Type 303 stainless steel and can accommodate vessels up to 45.7 cm (18") in diameter within the sturdy U-shaped base.
Expand 6 Items
Loading...
VWR® Talon® Adjustable Cylinder Stand
VWR® Talon® Adjustable Cylinder Stand
Supplier: VWR International
Heavy-duty, cast aluminum stand with adjustable L-brackets holds cylinders from 15.2 – 23.5 cm (6 – 91/4") in diameter.
Expand 1 Items
Loading...
VWR® Talon® Heavy-Duty Support Stands
VWR® Talon® Heavy-Duty Support Stands
Supplier: VWR International
Heavy-duty support stands permit cylindrical vessels up to 30.5 cm (12") in diameter to sit close to the support rod.
Expand 6 Items
Loading...
High-Low Alcohol Burner Stand
High-Low Alcohol Burner Stand
Supplier: VWR International
High-Low Alcohol Burner Stand
Expand 2 Items
Loading...
Tripod Bases
Supplier: VWR International
These heavy cast iron bases provide a three-point support for stability.
Expand 3 Items
Loading...
Fun Stand
Fun Stand
Supplier: VWR International
Ensure accuracy in all your experiments.
Expand 1 Items
Loading...
Funnel Support with Base
Funnel Support with Base
Supplier: VWR International
Made from fine, oil finished hardwood designed to support up to four funnels. The height is adjustable and the holes are slotted for easy placement of funnels. The base measures 11 x 38 cm.
Expand 1 Items
Loading...
VWR® Talon® Aluminum Support Plate
VWR® Talon® Aluminum Support Plate
Supplier: VWR International
Talon® Aluminum Support Plate fits standard lattice lab frames using VWR® Talon® connectors 21572-501, 21572-556, or 60097-055.
Expand 1 Items
Loading...
T-Nesting Support Bases
Supplier: VWR International
Specially designed to provide excellent stability, these bases are shaped with legs that slide freely under one another for compact storage or set up.
Expand 1 Items
Loading...
VWR® Talon® Cylinder Stand
VWR® Talon® Cylinder Stand
Supplier: VWR International
Cast aluminum stand hinges open and has four thumbscrews to tightly grip cylinders from 20.3 – 22.9 cm (8 – 9") in diameter.
Expand 1 Items
Loading...
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
Supplier: VWR International
Support stand bases are constructed of cast iron with black enamel finish.
Expand 4 Items
Loading...
VWR® Talon® Support Stand, Cast Iron, Tripod Base
VWR® Talon® Support Stand, Cast Iron, Tripod Base
Supplier: VWR International
Support stand base is constructed of cast iron with black enamel finish.
Expand 1 Items
Loading...
VWR® Talon® Double Buret Holder and Support Stand
VWR® Talon® Double Buret Holder and Support Stand
Supplier: VWR International
Double buret clamp holds any size buret from micro to 100 mL capacity.
Expand 6 Items
Loading...
Buret Holder
Buret Holder
Supplier: Avantor
Developed by a Lab Technician.
Expand 1 Items
Loading...
Labjaws FootPlate, Ace Glass Incorporated
Labjaws FootPlate, Ace Glass Incorporated
Supplier: Ace Glass
The Labjaws Footplate holds Flexaframe poles
Expand 1 Items
Loading...
Bench Top Reactor Stand, Ace Glass Incorporated
Bench Top Reactor Stand, Ace Glass Incorporated
Supplier: Ace Glass
This newer bench reactor stand assembly accommodates all types of glass bench filter and pressure reactors.
Expand 1 Items
Loading...
SP Bel-Art Compact Support Stand, Bel-Art Products, a part of SP
SP Bel-Art Compact Support Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This non-corrosive support stand with stainless steel mounting rod provides modern advantages over metal ring stands.
Expand 1 Items
Loading...
LaboPlast Funnel Stands for Two or Four Funnels, Bürkle
LaboPlast Funnel Stands for Two or Four Funnels, Bürkle
Supplier: BURKLE INC
Sturdy and stable funnel stand, resistant to chemicals, shatter-proof and with continuously variable height adjustment.
Expand 2 Items
Loading...
SP Bel-Art Economy Support Stand, Bel-Art Products, a part of SP
SP Bel-Art Economy Support Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Sturdy and stable molded polycarbonate base supports a 45.7 cm (18") tall stainless steel support rod.
Expand 1 Items
Loading...
Photochemical Reactor Stand and Stand Inserts, Ace Glass Incorporated
Photochemical Reactor Stand and Stand Inserts, Ace Glass Incorporated
Supplier: Ace Glass
Sturdy aluminum, powder coated stand for use with cylindrical reactors.
Expand 9 Items
Loading...
Plate Support Shelf, Ace Glass
Plate Support Shelf, Ace Glass
Supplier: Ace Glass
Adjustable cast alloy, black epoxy coated support shelf
Expand 2 Items
Loading...
Support Stand, 'C' Shape, All Stainless Steel, Ace Glass Incorporated
Support Stand, 'C' Shape, All Stainless Steel, Ace Glass Incorporated
Supplier: Ace Glass
An all-stainless-steel support stand with a heavy, six pound base
Expand 2 Items
Loading...
Sort by
Recommended for You