ICP-MS Internal Standard for 6020, Specpure®, Thermo Scientific
Catalog # CAAA43161-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:100 ml
- Cat. no.:CAAA43161-AE
- Supplier No.:43161-AE
Specifications
About this item
Molecular Formula: Matrix: 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD01868829