ICP-MS Tuning Standard solution for HP®, Specpure®, Thermo Scientific
CAAA42586-AE
:
- Size:100 ml
- Cat. no.:CAAA42586-AE
- Supplier No.:42586-AE
Molecular Formula: Matrix: 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00240453
Molecular Formula: Matrix: 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00240453