Multi-Element QC-21 Check Standard Solution, Specpure®, Thermo Scientific
Molecular Formula: Matrix: 5% HNO3/tr. F-/tr. tartaric acid
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00803610
Frequently Bought Together
Molecular Formula: Matrix: 5% HNO3/tr. F-/tr. tartaric acid
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00803610