ICP-MS Stock Standard solution B for 200.8, Specpure®, Thermo Scientific
CAAA40486-AE
:
- Size:100 ml
- Cat. no.:CAAA40486-AE
- Supplier No.:40486-AE
Molecular Formula: Matrix: 1% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00803327
Molecular Formula: Matrix: 1% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00803327