Multi-Element QC-7 Check Standard Solution, Specpure®, Thermo Scientific
About this item
Molecular Formula: Matrix: 5% HNO3/tr. F-
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00210654
1 Options Available Below
Product Details & Documents
Molecular Formula: Matrix: 5% HNO3/tr. F-
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00210654
Documents
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: 39145-AE
- 500 mlDiscontinued-Supplier Number: -
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



