You searched for: Filter Plates
Essential for biomolecule purifications and separations filter plates are offered in different filtration media membrane to accommodate any array applications. With individual membranes and fluid directors within each cavity, the cross contamination between wells is eliminated. Filter plates are specifically designed for a clog-free liquid flow completed at a fast and uniform rate for higher yields. Common well formats are compatible for use with standard processing equipment.
AcroPrep™ 96-Well Filter Plates for Solvent Filtration, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroPrep Advance 96-well filter plates are chemically compatible with harsh organic and / or aqueous solvents and contain inert materials of construction to provide a stable platform and ensure complete recovery of samples and low non-specific binding. The water wettable Polytetrafluoroethylene (wwPTFE) is a universally chemically compatible membrane and offers fast flow rates, low protein binding, and low levels of UV-absorbing extractables. These plates are suitable for applications such as metabolic studies, molecular or drug synthesis reactions, sample prep using solvents, and aggressive filtration applications.
Expand 18 Items
AcroPrep™ 24-Well Filter Plates, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The Acroprep 24-well filter plate utilizes Pall’s proprietary high-performance multi-layer filter media and membranes to offer time savings, strong performance claims and streamlined workflow improvements in a 24-well plate format.
Expand 28 Items
AcroPrep™ Advance 96-Well Filter Plates, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroPrep™ Advance Filter Plates feature polypropylene plate housing, polystyrene lid, and borosilicate glass filter media.
Expand 1 Items
AcroPrep™ Filter Plates for Aqueous Filtration, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroPrep and AcroPrep Advance filter plates combine high performance membranes with unique well designs to provide fast processing with efficient removal of particulates. These plates are suitable for applications such as general sample prep, gross fractionation, call harvesting, and cell-based assays.
Expand 18 Items
AcroPrep™ Advance 96-Well Long Tip Filter Plates for Nucleic Acid Binding (NAB), Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The AcroPrep™ advance 96-well long tip filter plate for Nucleic Acid Binding (NAB plate) incorporates a silica-based quartz glass fiber media to allow for efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples.
Expand 1 Items
AcroPrep™ Advance Filter Plates for Lysate Clearance, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroPrep™ Advance Filter Plates for Lysate Clearance are manufactured with biologically inert materials that allow clarification of most types of lysates without loss of target molecules.
Expand 4 Items
AcroPrep™ Filter Plates For Ultrafiltration, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
AcroPrep and AcroPrep Advance filter plates combine high performance Omega membranes with unique well designs to provide rapid, efficient separation of biomolecules. These plates are suitable for applications such as size exclusion, PCR clean-up, nucleic acid purification, and protein separation.
Expand 14 Items
AcroPrep™ Advance 96-Well Filter Plates for Nucleic Acid Purification, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These plates are for maximum yields of high quality DNA.
Expand 2 Items
AcroPrep™ Advance Protein Purification 96-Well Filter Plates, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications.
Expand 5 Items
AcroPrep™ 96-Well Filter Plates, 1 ml, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Constructed of chemically resistant, biologically inert polypropylene, plates are optimized for a variety of applications requiring larger per well volume processing.
Expand 5 Items
Corning® FiltrEX™ 96-Well Filter Plates, Corning
Supplier: Corning
FiltrEX™ filter plates meet the industry standards for plate dimensions. The rigid side walls make the plate ideal for automation and the wide skirt accepts barcodes. Individual filter discs are encapsulated in the plate by a process that ensures 100% integrity of each well. The design of the nozzle prevents sample wicking and cross-contamination. Glass fibre filter plates can be used for a variety of applications, such as plasmid isolation, DNA purification, PCR clean-up or receptor/ligand binding assays. The low-binding hydrophilic PVDF membrane can be used for lysate clarification, protein kinase assays, or bead- or resin-based separation assays.
Expand 5 Items
Corning® BioCoat™ Pre-Coated PAMPA Plate System, Corning
Supplier: Corning
A 96-well, pre-coated filter plate, individually packaged with a 96-well microassay receiver plate.
Expand 1 Items
Captiva Filter Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Captiva dual-depth filtration media removes precipitated proteins and prevents HPLC column clogging.
Expand 9 Items
Captiva ND and ND Lipids Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Captiva ND enables in-well protein precipitation; Captiva ND Lipids removes biofluid phospholipids.
Expand 3 Items
GST MultiTrap Filter Plates, Cytiva
Supplier: Cytiva
GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and purification of glutathione S-transferase (GST) tagged proteins
Expand 1 Items
His MultiTrap TALON Filter Plates, Cytiva
Supplier: Cytiva
His MultiTrap™ TALON® multiwell plates are designed for high-throughput screening of histidine-tagged proteins from clarified cell lysates. His MultiTrap™ TALON® prepacked plates are manufactured with high reproducibility with regards to the well and media volumes in order to enable consistent results. The average purity of His MultiTrap TALON® is >85% with yields up to 1 mg/well. Leakage of cobalt ions from TALON® Superflow™ medium is negligible. His MultiTrap™ TALON® is used manually with vacuum or centrifugation, or in a liquid handling station.
Expand 1 Items
His MultiTrap FF and HP Filter Plates, Cytiva
Supplier: Cytiva
His MultiTrap FF and His MultiTrap HP are prepacked disposable 96-well plates for reproducible high throughput parallel purification of histidine-tagged recombinant proteins by Immobilized Metal ion Affinity Chromatography (IMAC).
Expand 2 Items
Filter Plates, 96-Well, Protein A HP MultiTrap™, Cytiva
Supplier: Cytiva
Protein A Sepharose High Performance has high affinity for the Fc region of IgG from a variety of species.
Expand 1 Items
PD MultiTrap™ G-25, Filter Plates, 96-Well, Cytiva
Supplier: Cytiva
PD MultiTrap G-25 is designed for high-throughput desalting and buffer exchange of multiple samples.
Expand 1 Items
Whatman™ UNIFILTER® Microplates, 384-Well, Whatman products (Cytiva)
Supplier: Cytiva
Cytiva's Whatman UNIFILTER 384 well microplates with filter bottom wells are used in DNA template clean-up, cell capture, removing unwanted debris from sample and reagents, and removing dye blobs in biological samples.
Expand 3 Items
Streptavidin HP MultiTrap™, Cytiva
Supplier: Cytiva
Streptavidin HP MultiTrap can be used for protein enrichment, where a biotinylated antibody (or similar affinity molecule) is attached to the Streptavidin and the protein of interest is enriched through the affinity interaction with the antibody.
Expand 1 Items
Protein G HP MultiTrap™, 96-Well, Cytiva
Supplier: Cytiva
Protein G Sepharose HP has high affinity for the Fc region of IgG from a variety of species.
Expand 1 Items
Whatman™ 850-DS 8-Channel Filter Plates, Whatman products (Cytiva)
Supplier: Cytiva
Whatman 8-channel filter plates are specifically designed for use with the Agilent™ 850-DS Dissolution Sampling Station.
Expand 9 Items
Nunc® 96-Well Filter Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
Filter plates are made of polypropylene, which offers chemical resistance and mechanical strength
Expand 3 Items
Homogenizer Plates, E-Z 96®, and Spin Columns, Omega Bio-Tek
Supplier: Omega Bio-Tek
For simple and rapid homogenisation of cell and tissue lysates.
Expand 3 Items
Whatman™ UNIFILTER® Microplates, 24-Well, Whatman products (Cytiva)
Supplier: Cytiva
The 10 ml UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.
Expand 3 Items
Whatman™ UNIFILTER® Microplates, 96-Well, Whatman products (Cytiva)
Supplier: Cytiva
UNIFILTER 96-well microplate, 800 µl, long drip director.
Expand 7 Items
Pierce™ HisPur™ Cobalt Spin Filter Plates, 96 Well, Thermo Scientific
Supplier: Thermo Scientific
HisPur™ Cobalt spin plates are convenient, ready-to-use, pre-dispensed filter plates for the efficient purification of polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells
Expand 1 Items
Pierce™ Detergent Removal Spin Plates, Thermo Scientific
Supplier: Thermo Scientific
Pierce™ Detergent removal filter spin plates enable multiple-sample processing using a high-performance resin that offers effective detergent removal and excellent protein/peptide recovery.
Expand 1 Items
Pierce™ Zeba™ 96-well Spin Desalting Plates, Thermo Scientific
Supplier: Thermo Scientific
Zeba™ 96-well Spin Desalting Plates (40K MWCO) contain a size-exclusion chromatography resin that provides excellent protein desalting performance in a centrifuge format for 5 µL to 4 mL samples.