NeuLog WiFi Module
Neulog WiFi 2.0
Catalog # 470221-194
Supplier: Wards
Supplier #: WIFI202
Jump to:
Product Details & Documents
Wi-Fi simplifies data acquisition.
- Compatible with iPads, tablets, and PCs
- No downloads or installations
- Compatible with popular browsers
The Wi-Fi module is one of the most advanced and innovative elements of NeuLog. Using Wi-Fi technology, students can take measurements with NeuLog on nearly any kind of device including a tablet with Android, a PC computer with Windows or Linux, or a Macintosh with OS.
These devices communicate with the Wi-Fi adaptor for a unique experience of data acquisition. Users don’t need to download any applications or install any software to use NeuLog sensors with the Wi-Fi module. Any popular browser program can be used.
Caution: For educational use only.
Documents
TDS NeuLog WiFi Module.pdf
Datasheet
Specifications
- Description:Neulog WiFi 2.0
- Cat. No.:470221-194
- Interface:WiFi