Stirrer Magnets, Triangular, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
PTFE "V" shaped vane with a mounted magnetic bar cross
Stirrer magnets are designed for use with certain Microscale vials. They can also can be used with any V-bottom reaction vials or tubes. Stirrer magnet 89078-004 is designed for use with a 6–7mm tube I.D. Stirrer magnet 89078-006 is designed for use with a 9–10mm tube I.D. Stirrer magnet 89078-008 is designed for use with a 11–12mm tube I.D.
Frequently Bought Together