To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Bovine
- Antigen symbol:IgG
- Conjugation:RPE (R-Phycoerythrin)
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:500 µg
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Antigen synonyms:F(ab')2 Anti-Bovine IgG (H&L)
- Antibody fragment:F(ab')2
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide, 10 mg/ml Polyethylene Glycol (PEG-8000)
- Storage temperature:+4 °C
- Concentration:0.5 mg/ml by absorbance = 82.0 at 565 nm
- Shipping temperature:Ambient
- Immunogen:Bovine IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:RL701-408-002
- Supplier No.:701-408-002
Specifications
About this item
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Bovine