Order Entry
ContactUsLinkComponent
Anti-IgG Rabbit Polyclonal Antibody
Anti-IgG Rabbit Polyclonal Antibody
Catalog #: RL701-401-002
Anti-IgG Rabbit Polyclonal Antibody
Catalog #: RL701-401-002
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Bovine
  • Antigen symbol:
    IgG
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    1 mg
  • Western blot:
    Yes
  • Epitope:
    Heavy and Light chain
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Antigen synonyms:
    F(ab')2 Anti-Bovine IgG (H&L)
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
  • Storage temperature:
    –20 °C
  • Concentration:
    1.0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Wet Ice
  • Immunogen:
    Bovine IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Cat. no.:
    RL701-401-002
  • Supplier No.:
    701-401-002

Specifications

About this item

Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:20000-1:300,000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Bovine

Customers Who Bought This Also Bought