Anti-CLVS1 Rabbit Polyclonal Antibody
Catalog # RL612-401-D17
Supplier: Rockland Immunochemical
Specifications
- Antibody type:Primary
- Antigen name:clavesin 1
- Antigen symbol:CLVS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Size:100 µL
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:366311
- Antigen synonyms:RGD1564200|Rlbp1l1|Clavesin-1
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Clavesin Antibody was produced in rabbit by repeated immunizations with synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH.
- Purification:affinity-purified
- Cat. no.:RL612-401-D17
- Supplier No.:612-401-D17
Specifications
About this item
Anti-Clavesin antibody is suitable for use in ELISA, Western Blotting and ICC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 35 kDa in size corresponding to clavesin Protein doublet in Western Blots.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry 1:50
Type: Primary
Antigen: CLVS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: