Orders LinkContactUsLinkComponent
Solid Polyethylene Acid Cabinets, Justrite®
Solid Polyethylene Acid Cabinets, Justrite®
Supplier:   Justrite
Solid Polyethylene Acid Cabinets, Justrite®
Supplier:  Justrite
Certificates
Solid Polyethylene Acid Cabinets, Justrite®
Solid Polyethylene Acid Cabinets, Justrite®
3 Options Available Below
Product Details & Documents
These solid wall high density polyethylene cabinets have excellent chemical resistance to handle the most corrosive chemicals.
  • Sturdy, smooth construction in an attractive design
  • Suitable for highly aggressive liquids - no metal parts to corrode
  • Three-point door latch on larger units offer secure closure
  • Toe kick on undercounter model allows user to stand closer to counter - less leaning, better ergonomics
They are 100% poly welded with no metal parts, they include a external 2" NPT vent/exhaust connections and have padlockable hasps for security. White.
Documents
Product Family Options
Product Information
  • Capacity
    Material
    Availability
    Price
  • 97048-664
    Acids benchtop cabinet
    Acids benchtop cabinet
    1 door, countertop, 6× 2½ L bottles
    Polyethylene
    Log in for availability
    Log in for availability
    Each
    Each - $14,791.29
    $14,791.29
    Each
       $14,791.29
    Supplier Number: 24004
  • 97048-668
    Acids stand-alone cabinet
    Acids stand-alone cabinet
    2 door, stand alone, 36× 2½ L bottles
    Polyethylene
    Log in for availability
    Log in for availability
    Each
    Each - $17,884.76
    $17,884.76
    Each
       $17,884.76
    Supplier Number: 24015
  • 97048-666
    Acids underbench cabinet
    Acids underbench cabinet
    2 door, undercounter, 36× 2½ L bottles
    Polyethylene
    Log in for availability
    Log in for availability
    Each
    Each - $16,436.27
    $16,436.27
    Each
       $16,436.27
    Supplier Number: 24010