Plier/Cutters, Forming Dog Leg with Shear Cut, Excelta
Catalog # 63042-422
Supplier: Excelta
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Five Star Quality Plier/Cutters
- Material:Carbon steel
- Cat. no.:63042-422
- Cutting Capacity:0.025" (max.)
- Tip Style:Shear Cut Type
- Overall Length:5¼"
- Supplier No.:500-89-US
Specifications
About this item
This design will form and cut 'Dog Leg' in one operation and can be used for surface mount or through hole boards.