ContactUsLinkComponent
Ultimate FX Full Facepiece Air Purifying Respirator FF-400, 3M
Ultimate FX Full Facepiece Air Purifying Respirator FF-400, 3M
  89135-090
 :  3M
undefined
Ultimate FX Full Facepiece Air Purifying Respirator FF-400, 3M
  89135-090
 :  3M
 :  7000002284
  • Size:
    Medium
  • Cat. no.:
    89135-090
  • Supplier No.:
    FF-402
Designed for flexibility in extreme environments.
  • Head harness, comfort cradle, and soft silicone nose cup allow secure, comfortable wear over extended periods
  • Large lens provides superior peripheral vision
  • Bonded silicone gaskets and easy-to-use respirator cartridge/filter attachments
Scotchgard™ protector helps lens resist dirt, paint, and stains. Cool Flow™ exhalation valve reduces heat and moisture buildup inside facepiece.