Order Entry
Canada
ContactUsLinkComponent
SP Bel-Art Spill Containment Tray, Bel-Art Products, a part of SP
SP Bel-Art Spill Containment Tray, Bel-Art Products, a part of SP
  58948-298
undefined
SP Bel-Art Spill Containment Tray, Bel-Art Products, a part of SP
  58948-298
 :  F24675-0050

 

  • Cat. no.:
    58948-298
  • W×L:
    69 x 58 cm (27 x 23")
  • Supplier No.:
    246750050

 

 

The polystyrene tray accepts 47727-664 Labmat™ Sheets as liners (not included) to absorb spilled liquids.

  • Catch spills when containers leak, spill, or crack
  • Ideal for storing small containers on lab countertop
  • Chemical-resistant polyethylene trays have built-in containment reservoir
  • Translucent sidewalls make leaks visible
  • Pour spout allows easy draining

The tray is 12.7 mm (½") deep with an external flange.