Hand Sink, with Knee/Foot Valve, Eagle MHC
Catalog # 71001-942
Supplier: Eagle Group
Specifications
- Bowl Dimensions (W×L×H):9¾×13½×6¾"
- Width:37.46 cm (14¾")
- Height:17½"
- Length:18⅞"
- Cat. no.:71001-942
- Supplier No.:HSA-10-1FK
Specifications
About this item
Hand sink is made up of heavy gauge type 304 stainless steel.
- Inverted 'V' edge to prevent spillage, and splash mounted gooseneck spout
- Deep-drawn bowl with positive drainage
- All-welded construction
Delivery: Includes single knee pedal and stainless steel skirt and basket drain.