Mantle, Fabric/Cloth, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Designed for use with certain reaction flasks, such as 89054-900 and 89055-058
Upper temperature limit is 450°C (842°F). Mantles must be operated through a temperature controller.
Certifications: CSA approved.
Ordering information: Supplied with detachable 121.9cm (4') cord and locking connector.
Customers Who Bought This Also Bought