To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glycoprotein Ib (platelet), Alpha Polypeptide
- Antigen symbol:GP1BA
- Clonality:Monoclonal
- Clone:SZ2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:2811
- Antigen synonyms:GP1B|CD42b-alpha|CD42B|BSS|VWDP|BDPLT3|GPIbA|BDPLT1|DBPLT3
- Storage buffer:Phosphate-buffered saline, pH 7.2 containing 1mg/ml BSA and 0.1% sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.2 mg/ml
- Immunogen:Tissue / cell preparation: Human washed platelets.
- Tested applications:Fluorescence-Activated Cell Sorting
- Purification:Protein A affinity purified
- Cat. no.:89360-658
- Supplier No.:GTX19655
Specifications
About this item
CD42b is a platelet activation marker involved in the process of coagulation as an aggregating factor. It interacts with extracellular matrix as well as with adhesion molecules. Its expression and activation seems to be regulated by VEGF and PDGF.
For FACS: Use 2?g for 5x10^6 cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GP1BA
Clonality: Monoclonal
Clone: SZ2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human