Order Entry
Canada
ContactUsLinkComponent
Anti-Hepatitis C NS4 Mouse Monoclonal Antibody
Anti-Hepatitis C NS4 Mouse Monoclonal Antibody
  89360-544
 :  Genetex
Anti-Hepatitis C NS4 Mouse Monoclonal Antibody
  89360-544
 :  Genetex
 :  GTX19048

 

  • Antibody type:
    Primary
  • Antigen name:
    Hepatitis C NS4
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage buffer:
    Preservative: None. Constituents: 0.01M PBS. pH 7.0
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide (Human) in which overlapping sequences of at least 90 residues in length were selected from the immunodominant NS4 regions of Chinese HCV strains.
  • Tested applications:
    ELISA, IHC-P
  • Purification:
    Protein G purified
  • Cat. no.:
    89360-544
  • Supplier No.:
    GTX19048

 

 

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.

Type: Primary
Antigen: Hepatitis C NS4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human