Anti-STK25 Rabbit Polyclonal Antibody
89355-036
: Genetex
- Antibody type:Primary
- Antigen name:Serine/Threonine Kinase 25
- Antigen symbol:STK25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:10494
- Antigen synonyms:Ste20-like kinase|SOK-1|YSK1|Sterile 20/oxidant stress-response kinase 1|Ste20-like|SOK1
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:48 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.38mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human STK25. The exact sequence is proprietary.
- Tested applications:ICC/IF, IHC-P, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89355-036
- Supplier No.:GTX107634
Rabbit Polyclonal antibody to STK25 (serine/threonine kinase 25 (STE20 homolog, yeast))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: STK25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human