Anti-PSKH1 Rabbit Polyclonal Antibody
Catalog # 89355-026
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Protein Serine Kinase H
- Antigen symbol:PSKH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:5681
- Antigen synonyms:PSK-H1|Serine/threonine-protein kinase H1|PSKH1
- Storage buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular weight:48 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.72mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human PSKH1. The exact sequence is proprietary.
- Tested applications:ICC/IF, WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89355-026
- Supplier No.:GTX107585
Specifications
About this item
Rabbit polyclonal antibody to PSKH1 (protein serine kinase H1)
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PSKH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human