Anti-MTOR Rabbit Polyclonal Antibody
89349-122
: Genetex
- Antibody type:Primary
- Antigen name:Mechanistic Target of Rapamycin
- Antigen symbol:MTOR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- ChIP:Yes
- Gene ID:2475
- Antigen synonyms:FRAP1|mTOR|m-TOR|RAPT1|RAFT1|FKBP-Rapamycin Associated Protein|Mechanistic Target Of Rapamycin|EC 2.7.11.1|FRAP|FRAP2|Rapamycin Target Protein|FK506 binding protein 12-rapamycin associated protein|Mammalian Target Of Rapamycin|Serine/Threonine-Protein Kinase MTOR|MTOR
- Storage buffer:1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.
- Molecular weight:289 kDa
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the C-terminus region of human mTOR. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89349-122
- Supplier No.:GTX101557
Rabbit Polyclonal antibody to mTOR (mechanistic target of rapamycin (serine/threonine kinase))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IP: 1:100-1:500, ChIP. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human