Optics Bench Kit
Optics Bench Kit
Catalog # 470137-548
Supplier: Wards
Supplier #: PH0649
Jump to:
Product Details & Documents
Study optics with this complete kit while learning about light, refraction, reflection, convex and concave lenses, and focal length.
This enhanced set of accessories allows users to perform a wide variety of experiments in optics.
: Power supply required; not included.
: The kit includes: a bench, riders with tightening screws, a lamp with halogen bulb, lens holders, a variety of lenses, set of diaphragms, a mirror, and various screens.
Specifications
- Description:Optics Bench Kit