Order Entry
Canada
ContactUsLinkComponent
Mag-Bind® Total RNA and Total RNA 96 Kits, Omega Bio-tek®
Mag-Bind® Total RNA and Total RNA 96 Kits, Omega Bio-tek®
Catalog # CA89167-360
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Total RNA and Total RNA 96 Kits, Omega Bio-tek®
Catalog # CA89167-360
Supplier:  Omega Bio-Tek
Supplier Number:  M6731-01
CAS Number:  

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Mag-Bind® Total RNA 96 Kit
  • Size:
    4 x 96 Preps
  • Description:
    Mag-Bind® Total RNA 96 Kit
  • Cat. no.:
    CA89167-360
  • Includes:
    Mag-Bind® Particles Solution R, Buffer RCL, Buffer PNP, Mag-Bind® Particles LC, RWB, LPA Buffer, DEPC Water
  • Supplier No.:
    M6731-01

Specifications

About this item

The Mag-Bind Total® RNA Kit and Mag-Bind® Total RNA 96 Kit both provide a novel technology for total RNA isolation

These kits allow the rapid and reliable isolation of high-quality, total cellular RNA from a wide variety of cells and tissues. The system combines the higher nucleic acid-binding properties of Mag-Bind® Particles with the E.Z.N.A.® RNA Isolation system to provide a superior quality of RNA. Unlike column-based systems, the binding of nucleic acids to magnetic particles occurs in solution, resulting in increased binding kinetics and binding efficiency. Particles are also completely re-suspended during the wash steps of the purification protocol, which enhances contact with the wash buffer and removal of contaminants, thus increasing nucleic acid purity. The E.Z.N.A.® Mag-Bind® Total RNA procedure can be fully automated with most robotic workstations.