About this item
The E.Z.N.A.® HP total RNA isolation kit provides a rapid and easy method for RNA isolation from a small amount of cultured eukaryotic cells or tissues
- High Purity
This kit allows single or simultaneous processing of multiple samples in less than 40 minutes. Normally, 1×10^7 eukaryotic cells or 25 to 30 mg tissue can be used in a single experiment. There is no need for phenol/chloroform extractions, time-consuming steps such as CsCl gradient ultracentrifugation, or precipitation with isopropanol or LiCl. RNA purified using the E.Z.N.A.® HP Total RNA method is ready for applications such as RT-PCR, RT-qPCR, Northern blotting, poly A+ RNA (mRNA) purification, nuclease protection, and in vitro translation.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Description:HP Total RNA Kit