

Acetonitrile-[D₃] ≥99% (Isotopic), MagniSolv™ for NMR spectroscopy, Sigma-Aldrich®
Catalog # CA1.02904.0010
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle, septum-sealed
- Size:10 mL
- Cat. no.:CA1.02904.0010
- Supplier No.:1.02904.0010
Specifications
About this item
:
:44.03 g/mol
Boiling Pt:80.7 °C (1013 hPa)
Melting Pt:–42 °C
Density:0.844 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-616-5
UN:1648
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881