Eyesaver Drench Hose and Eyewash Combination, Model SE-925, Speakman®
Catalog #: VF-0925-SE
Supplier: Speakman
Small Business Enterprise
Specifications
- Description:Eyesaver Drench Hose and Eyewash Combination, Model SE-925
- Material:Stainless Steel, Brass, Plastic, and Steel Composition
- Cat. no.:VF-0925-SE
- Finish:Polished Chrome and Yellow
- Supplier No.:SE-925
Specifications
About this item
Speakman® Drench hose and eyewash combinations provide great flexibility in laboratories and other industrial areas for flushing the eyes and body.
- Features a stay-open valve allowing hands-free operation oce the eyewash has been activated
All units come standard with a flexible hose making an ideal product for direct flushing on contaminated areas. Model SE-925 is a wall mounted drench hose/eyewash featuring a 12' recoiling hose and wall bracket.
Certifications: Meets ANSI 358.1.
Customers Who Bought This Also Bought