ContactUsLinkComponent
Safe-T-Zone® Select Series Combination Shower, Model SE-616, Speakman®
Safe-T-Zone® Select Series Combination Shower, Model SE-616, Speakman®
Catalog # VF-SE-616
Supplier:  Speakman
Small Business Enterprise
undefined
Safe-T-Zone® Select Series Combination Shower, Model SE-616, Speakman®
Catalog # VF-SE-616
Supplier:  Speakman
Supplier Number:  SE-616
Small Business Enterprise
Specifications
  • Description:
    Safe-T-Zone® Select Series Combination Shower, Model SE-616
  • Material:
    Stainless Steel, Brass, Plastic, and Steel Composition
  • Cat. no.:
    VF-SE-616
  • Inlet Size:
    1.25" NPT
  • Supplier No.:
    SE-616
Specifications
About this item
Speakman® Safe-T-Zone® Select Series combination shower and eyewash unit features a combination eyewash with two yellow aerated nylon spray outlets and a rectangular bowl.
  • Showerhead is activated by a high visibility yellow pull rod
The outlets and bowl are combined on a DuraJade™ powder-coated, galvanized steel stanchion with an impeller action deluge showerhead. The Model SE-616 combination shower is supplied with an SE-505 eyewash with stainless steel bowl.
Certifications: Meets ANSI 358.1.