Tetrabromofluorescein potassium salt ≥85.0% (by HPLC)
Catalogue # TCT0036-025G
Fournisseur: TCI America
Numéro CAS:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Size:25 g
- Cat. no.:TCT0036-025G
- Supplier No.:T0036-25G
Spécifications
Résultats des tests de spécification
- Purity/Analysis Method:85.0% (HPLC)
Résultats des tests de spécification
À propos de cet article
Numéro CAS:56897-54-2
:
Formule:C₂₀H₈Br₄O₅
Numéro MDL:MFCD00070510
:724.08 g/mol