You searched for: Microcentrifuges
Compact microcentrifuges isolate DNA or simply separate micro volume mixtures. Load microtubes and standard or dell-welled plates in the instruments easily. Adaptable to any industrial or laboratory setting, non-refrigerated or refrigerated models come with multiple mode options for the high-speed and powerful rotors. Continuous or pulse spinning options move samples fast and consistently while included timers alert with sample preparation is complete. Microcentrifuges may come with or without the protective lid.
VWR® Mini Centrifuges
Supplier: VWR International
The VWR® Mini Centrifuge is designed for quick spin-downs of microcentrifuge tubes and PCR strips and tubes.
Expand 1 Items
VWR® High Speed Microcentrifuge
Supplier: VWR International
The compact, affordable VWR® High Speed Microcentrifuge is capable of high speed performance up to 15500 rpm/16100×g.
Expand 1 Items
VWR® Plate Centrifuge
Supplier: VWR International
Spins at 2500 rpm/ 850xg, to quickly spin down droplets and condensation in PCR plates.
Expand 1 Items
VWR® Compact Micro Centrifuges
Supplier: VWR International
These compact microcentrifuges are ideal for DNA, RNA, and temperature-sensitive samples.
Expand 9 Items
VWR® Micro Star 21 and 21R Microcentrifuges
Supplier: VWR International
VWR® Micro Star 21 ventilated and 21R refrigerated microcentrifuge with 24×1.5/2.0 ml rotor with ClickSeal biocontainment lid offer exceptional acceleration of up to 21000 xg in as little as 12 seconds.
Expand 2 Items
Hettich MIKRO 185 Microliter Centrifuges, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
Hettich Lab Technology's MIKRO 185 tabletop microliter centrifuge delivers application versatility and fast results in an elegant, compact frame.
Expand 3 Items
Stuart® 8500 rpm SCF4 Microcentrifuge, Cole Parmer
Supplier: Argos Technologies
The Stuart 8500 microcentrifuge provides high speed centrifugation with enhanced safety features.
Expand 1 Items
Microcentrifuges, 230 V, 5418/5418R
Supplier: Eppendorf
Ultra-compact, 18-place microcentrifuges are quiet and user-friendly
Expand 1 Items
Eppendorf 5427R Microcentrifuges
Supplier: Eppendorf
Labs demanding high sample throughput will benefit from the 5427R, featuring a double-row 48-place rotor to increase efficiency and improve running time.
Expand 1 Items
Hettich MIKRO 220 Microliter Centrifuges, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
The Hettich Lab Technology MIKRO 220 microliter centrifuge series offers large capacities and high-performance for high-throughput laboratories.
Expand 3 Items
Microcentrifuge, 5425, GLP, Eppendorf
Supplier: Eppendorf
Microcentrifuge 5425 plus the 1.5/2 ml kit rotor is the optimal choice for spin-column-based nucleic acid purification kits containing multiple centrifugation steps with frequent vessel changes, as used in target antigen production, sequencing sample preparation and antibody humanization.
Expand 12 Items
Microcentrifuges, 230 V, 5430 and 5430R
Supplier: Eppendorf
Microcentrifuges, 230 V, 5430 and 5430R
Expand 5 Items
Eppendorf® MiniSpin® Personal Microcentrifuges
Supplier: Eppendorf
Microcentrifuges are compact and easy to use, yet powerful enough to enable each workstation to be equipped with a “personal” centrifuge for a wide range of molecular biology separations and quick spins
Expand 2 Items
Microfuge® 16 Microcentrifuge, Beckman Coulter®
Supplier: Beckman Coulter
This non-refrigerated, cold-room compatible, high-performance microcentrifuge is ideal for fast pelleting or isolation of DNA, RNA, proteins, and viruses.
Expand 1 Items
mySPIN™ Mini Centrifuges Series, Thermo Scientific
Supplier: THERMO FISHER SCIENTIFIC MX
Bring increased efficiency and user comfort to your lab with mySPIN™ 6 and 12 mini centrifuges.
Expand 2 Items
Corning® LSE™ Mini Microcentrifuge, Corning
Supplier: Corning
The Corning® LSE™ mini microcentrifuge is a personal bench top instrument that operates at a fixed speed of 6000 min⁻¹ (2000×g) and was designed for quick spin downs of micro-samples.
Expand 1 Items
Eppendorf® Microcentrifuges, 5418/5418R
Supplier: Eppendorf
Ultra-compact, 18-place microcentrifuges are quiet and user-friendly
Expand 1 Items
Eppendorf® Microcentrifuges, Models 5430 and 5430R
Supplier: Eppendorf
Eppendorf; Includes aerosol-tight QuickLock rotor FA-45-16-17; High centrifugation speed up to 30130 x g (17500 rpm); Knob; 16x Eppendorf 5.0mL AT-QL rotor; 120 V, 50/60Hz.
Expand 17 Items
Hettich MIKRO 200 Microliter Centrifuges, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
The Hettich MIKRO 200 microliter centrifuge series is amongst the highest performing in its class.
Expand 2 Items
Microfuge® 20 Series IVD Microcentrifuges, Beckman Coulter®
Supplier: Beckman Coulter
Fast and versatile, the high-performance Microfuge® 20 Series are precisely designed to meet a wide variety of application needs.
Expand 6 Items
Eppendorf 5427R Centrifuges with Hydrocarbon Cooling
Supplier: Eppendorf
Eppendorf Centrifuge 5427 R supports your greener laboratory with hydrocarbon cooling. It is the perfect solution for laboratories with a variety of molecular biology applications or those in need of high-throughput solutions for 1.5 to 2 ml tubes - all combined in a microcentrifuge with the highest throughput/footprint-ratio within the Eppendorf microcentrifuge range.
Expand 3 Items
Eppendorf 5427R Centrifuges with Hydrocarbon Cooling, 230 V
Supplier: Eppendorf
Eppendorf Centrifuge 5427 R supports your greener laboratory with hydrocarbon cooling. It is the perfect solution for laboratories with a variety of molecular biology applications or those in need of high-throughput solutions for 1.5 to 2 ml tubes - all combined in a microcentrifuge with the highest throughput/footprint-ratio within the Eppendorf microcentrifuge range.
Expand 1 Items
Ohaus® Frontier™ 5000 Series Mini Centrifuge
Supplier: OHAUS CORP.
The Ohaus Frontier 5306 is a small but powerful minicentrifuge for simple and quick liquid separation.
Expand 1 Items
Cole-Parmer® CF-200 Series Stuart Programmable Microcentrifuge, Antylia Scientific
Supplier: Argos Technologies
Compact, powerful, and programmable.
Expand 1 Items
Labnet Prism R Refrigerated Microcentrifuge
Supplier: LABNET INTERNATIONAL SE
This high performance, refrigerated centrifuge can cool to 4°C in just eight minutes.
Expand 1 Items
VWR® High Speed Microcentrifuge, 220 V
Supplier: BENCHMARK SCIENTIFIC
The compact, affordable VWR® High Speed Microcentrifuge is capable of high speed performance up to 15500 rpm/16100×g.
Expand 1 Items
Mini Centrifuges, D1008, Dlab Scientific
Supplier: DLAB SCIENTIFIC INC.
The D1008 Mini centrifuge is ideal for microfiltration and quick spin-downs.
Expand 1 Items
Corning® LSE™ High Speed Microcentrifuge, Corning
Supplier: Corning
The Corning® LSE High Speed Microcentrifuge features an easy-to-use, digital control interface and high-speed performance for quick nucleic acid and protein separations
Expand 1 Items
SPROUT® Plus Mini Centrifuges
Supplier: Heathrow Scientific
Mini but mighty - ideal for small runs.
Expand 2 Items
HemataSTAT® II Centrifuge, Separation Technology
Supplier: Separation Technology
This microhematrocrit system provides a quantitative measure of the amount of red blood cells in relation to the amount of plasma