Order Entry
ContactUsLinkComponent
12594 results for Proteins and Peptides

You searched for: Proteins and Peptides

Proteins and Peptides

Proteins are used in routine laboratory procedures such as binding enzymes or coupling peptides to carrier proteins. These kits, mixture solutions, and collagen matrices fulfill a myriad of essential laboratory functions for developing relationships between proteins and other cellular components. The stimulating proteins offered have various amino acid arrangements and functions to fulfill any sample manipulation for testing purposes in any field.

Sort by
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Crystalline. Prepared by a cold alcohol isolation procedure. This salt-free preparation is the purest material ideal for sensitive biochemical and diagnostic assays. DNase- and RNase-free, virtually free of globulins and other interfering contaminants.

Expand 1 Items
Loading...
Leupeptin

Leupeptin

Supplier: VWR

A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin

Expand 2 Items
Loading...
Aprotinin

Aprotinin

Supplier: VWR

A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin

Expand 1 Items
Loading...
Leupeptin

Leupeptin

Supplier: VWR

Product is animal-free

Expand 1 Items
Loading...
Streptavidin

Streptavidin

Supplier: VWR

A 60 kDa tetrameric protein that exhibits high binding affinity for biotin. Able to bind one molecule of biotin with each subunit, streptavidin is useful in affinity chromatography, ELISA, immunohistochemistry and Western Blotting.

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Contains 0.1% sodium azide as preservative and strictly tested for RNase, DNase and Protease making it suitable for sensitive applications.

Expand 2 Items
Loading...
Casein

Casein

Supplier: VWR

High purity grade milk protein powder

Expand 1 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Bovine serum albumin, powder, Fraction V. Nuclease and protease-free. Suitable for use in most molecular biology applications.

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)

Bovine Serum Albumin (BSA)

Supplier: VWR

Concentrated BSA solution (Albumin, Bovine, Fraction V – Cold Alcohol Isolation) in 0.85% sodium chloride and contains 0.1% sodium azide (as a preservative for stability).

Expand 1 Items
Loading...
J.T.Baker®, Chicken Native Albumin (from Egg)

J.T.Baker®, Chicken Native Albumin (from Egg)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

CAS RN 9006-50-2. Powder, Purified. Packaged in a plastic container. 500g

Expand 2 Items
Loading...
Gelatin

Gelatin

Supplier: VWR

Gelatin

Expand 2 Items
Loading...
Bovine Serum Albumin (BSA)
Expand 2 Items
Loading...
Aprotinin

Aprotinin

Supplier: VWR

Product is animal-free

Expand 1 Items
Loading...
VWR Life Science Ribonuclease Inhibitor (RNase Inhibitor)

VWR Life Science Ribonuclease Inhibitor (RNase Inhibitor)

Supplier: VWR

RNase Inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro transcription, as well as for long term storage of valuable RNA samples

Expand 1 Items
Loading...
Pkcdelta Substrate

Pkcdelta Substrate

Supplier: Enzo Life Sciences

This peptide sequence, ARRKRKGSFFYGG, was selected from a peptide library for optimal phosphorylation by protein kinase C delta (PKCdelta).

Expand 1 Items
Loading...
Blocking Peptide for Heme Oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide

Blocking Peptide for Heme Oxygenase 1 pAb (Prod. No. BmL-HC3001) Blocking Peptide

Supplier: Enzo Life Sciences

Synthetic peptide corresponding to amino acid residues [12-25] + Cys-NH2 of human heme oxygenase-1. Complementary peptide to rabbit polyclonal antibody (BML-HC3001) for use in control experiments. Preincubation of antibody with this peptide abolishes the antibody reactivity with HO-1 protein.

Expand 1 Items
Loading...
LRRKtide, Biotin

LRRKtide, Biotin

Supplier: Enzo Life Sciences

LRRK substrate

Expand 1 Items
Loading...
Autocamtide-2-Related Inhibitory Peptide

Autocamtide-2-Related Inhibitory Peptide

Supplier: Enzo Life Sciences

CaM kinase II inhibitor

Expand 2 Items
Loading...
CDK2 Substrate

CDK2 Substrate

Supplier: Enzo Life Sciences

This peptide, HHASPRK, is a specific substrate for CDK2.

Expand 1 Items
Loading...
beta-Amyloid (1-40)

beta-Amyloid (1-40)

Supplier: Enzo Life Sciences

Expand 2 Items
Loading...
LRRKtide

LRRKtide

Supplier: Enzo Life Sciences

LRRK substrate

Expand 1 Items
Loading...
Mastoparan

Mastoparan

Supplier: Enzo Life Sciences

Mast cell degranulating peptide

Expand 1 Items
Loading...
Angiotensin I/II (1-7)

Angiotensin I/II (1-7)

Supplier: Enzo Life Sciences

Expand 1 Items
Loading...
Cyclosporin C

Cyclosporin C

Supplier: Enzo Life Sciences

Immunosuppressant

Expand 1 Items
Loading...
Substance P

Substance P

Supplier: Enzo Life Sciences

Neuropeptide

Expand 2 Items
Loading...
CdC2 Substrate

CdC2 Substrate

Supplier: Enzo Life Sciences

This peptide, (Ac-S)-PGRRRRK-biotin, is a substrate for CDC2 kinase.

Expand 1 Items
Loading...
IRS-1 (Tyr608) Peptide

IRS-1 (Tyr608) Peptide

Supplier: Enzo Life Sciences

Insulin receptor substrate

Expand 1 Items
Loading...
Bradykinin

Bradykinin

Supplier: Enzo Life Sciences

eNOS activator

Expand 2 Items
Loading...
Asialo-GM1

Asialo-GM1

Supplier: Enzo Life Sciences

Semisynthetic (from GM1).

Expand 2 Items
Loading...

GP antagonist-2

Supplier: Enzo Life Sciences

G protein antagonist

Expand 1 Items
Loading...
Sort by
Recommended for You