Order Entry
Canada
ContactUsLinkComponent
N,N-Dimethylformamide ≥99.5%
Flammable
Health
Irritant
N,N-Dimethylformamide ≥99.5%
Catalog # TCD0939-100ML
Supplier:  TCI America
CAS Number:  
Flammable
Health
Irritant
N,N-Dimethylformamide ≥99.5%
Catalog # TCD0939-100ML
Supplier:  TCI America
Supplier Number:  D0939-100ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 ml
  • Cat. no.:
    TCD0939-100ML
  • Supplier No.:
    D0939-100ML

Specifications

Specification Test Results

  • Purity/Analysis Method:
    99.5% (GC)

Specification Test Results

About this item

 :
 :73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C
Melting Pt:–61 °C
Density:0.95 (20 °C)
Flash Pt:60 °C
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284