NeuLog Photogate Logger Sensor
Catalog # 470137-386
Supplier: Wards
Specifications
- Resolution:100 µs
- Description:NeuLog photogate logger sensor
- Cat. no.:470137-386
- Accuracy:100 µs
- Supplier No.:NUL-209
Specifications
About this item
This sensor measures acceleration with one or two photogates and shows the status pictorially.
- Simple display features
- Four modes of operation: time, velocity, momentum, or acceleration
- Sensor displays values digitally and graphically
- Ideal for researching motion
Study motion via four modes of operation: time, velocity, momentum, or acceleration. You can select the modes of operation by clicking on their icon.
The sensor displays values digitally or graphically.
Ordering information: The following items are needed but not included: a battery module, viewer module (not needed if connecting to a computer to display data), USB module (not needed if using as a stand-alone probe or you have the Wi-Fi module), and Wi-Fi module (not needed if connecting to a computer through the USB module). A battery module and a USB module are recommended.
Caution: For educational use only.