NeuLog Turbidity Logger Sensor
ADC: 10 bit
Easily determine turbidity.
- Max. sample rate 100 S/sec.
- Experimental duration 1 sec. to 31 days
This sensor measures reflected light in order to determine solution turbidity. As the turbidity grows higher more light is reflected and measured in Nephelometric Turbidity Unit (NTU) by the light sensor which is located perpendicular to the tube.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module(Not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Documents
- Resolution:0.08 NTU
ADC: 10 bit - Description:NeuLog Turbidity Logger Sensor
- Range:0 - 200 NTU
- Cat. No.:470137-428
- Interface:USB