Orders LinkContactUsLinkComponent
NeuLog Wide-Range Temperature Logger Sensor, EISCO SCIENTIFIC LLC
NeuLog Wide-Range Temperature Logger Sensor, EISCO SCIENTIFIC LLC
NeuLog Wide-Range Temperature Logger Sensor, T: −200...1200 °C, T: 0.1 °C
ADC: 10 bit
Catalog # 470137-434
Supplier:   Wards
Supplier #:  NUL234
NeuLog Wide-Range Temperature Logger Sensor, EISCO SCIENTIFIC LLC
Catalog # 470137-434
Supplier:  Wards
NeuLog Wide-Range Temperature Logger Sensor, EISCO SCIENTIFIC LLC
NeuLog Wide-Range Temperature Logger Sensor, EISCO SCIENTIFIC LLC
NeuLog Wide-Range Temperature Logger Sensor, T: −200...1200 °C, T: 0.1 °C
ADC: 10 bit
Price
$187.30
Log in for availability
Each
Product Details & Documents
Measures a wide range of temperatures.
  • Max. sample rate 100 S/sec.
  • Experimental duration 1 sec. to 31 days
Based on a thermocouple sensor, this unit enables the measurement of very high and very low temperature levels. It runs in two modes of operation either Celsius or Fahrenheit.
Ordering information: For each probe you will need one or more of the following items: Battery Module, Viewer module (not needed if connecting to a computer to display data), USB module (not needed if only using as a stand alone probe or you have the WIFI module), WIFI module (Not needed if connecting to a computer through the USB module). Recommended is to have a battery module and a USB module.
Caution: For educational use only.
Specifications
  • Resolution:
    T: 0.1 °C
    ADC: 10 bit
  • Description:
    NeuLog Wide-Range Temperature Logger Sensor
  • Range:
    T: −200...1200 °C
  • Cat. No.:
    470137-434
  • Interface:
    USB