NeuLog Digital Display Module, EISCO SCIENTIFIC LLC
Catalog # 470137-436
Supplier: Avantor
Specifications
- Description:NeuLog viewer display module
- Cat. no.:470137-436
- Supplier No.:VIEW-200
Specifications
About this item
A simple display for use when working off-line.
- Connect to 1 or multiple NeuLog sensors
- 2 line digital display
The VIEW-200 is a small display module that can be connected to any chain of logger sensors working off-line with a Battery Module. The display module automatically searches for the connected sensors and displays one of them digitally. Scrolling the display of the reading of another sensor is done by pressing the push button on the module.
Ordering information: Requires 1 or more NeuLog sensors and a NeuLog Battery Module.
Caution: For educational use only.