Toluene saturated with water at 75 ° F, B S & W solvent
Catalog # RCSOD10-1
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. no.:RCSOD10-1
- Supplier No.:RSOD1000-4C
Specifications
Specification Test Results
- Appearance:Colorless liquid
Specification Test Results
About this item
:
:92.14 g/mol
Storage Temperature:Ambient
UN:1294
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512