Order Entry
Canada
ContactUsLinkComponent
Axygen® AxyPrep™ FragmentSelect-I Kit, Corning
Axygen® AxyPrep™ FragmentSelect-I Kit, Corning
Supplier:  Corning
Certificates
undefined
Axygen® AxyPrep™ FragmentSelect-I Kit, Corning
Supplier:  Corning
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    Sequencing reactions/libraries
  • Target molecule:
    DNA
  • Description:
    Axygen® AxyPrep™ FragmentSelect-I Kit

Specifications

Related Information

Frequently Bought Together