Order Entry
Canada
ContactUsLinkComponent
Axygen® AxyPrep™ Mag Tissue Genomic DNA Kit, Corning
Axygen® AxyPrep™ Mag Tissue Genomic DNA Kit, Corning
Supplier:  Corning
Certificates
Axygen® AxyPrep™ Mag Tissue Genomic DNA Kit, Corning
Supplier:  Corning
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    AxyPrep™ Mag Tissue Genomic DNA Kit

Specifications

Related Information

Frequently Bought Together