Highconic™ II Aluminum Fixed-Angle Rotor, Thermo Scientific
Thermo Scientific* HIGHConic II Aluminum Fixed-Angle Rotor delivers biocontainment at 10,350rpm.
Capacity flexibility with adapters ranging from 564 to 48mL. Choose from round or conical tubes with a wide array of available adapters.
: Biocontainment certified by CAMR*, HPA, Porton Down, U.K.
: Adapters are sold separately. Replacement sealing O-rings are also available (in sets of 2 with grease). Contact your Sales Representative for more information. Compatible with Thermo Scientific Megafuge 40/40R, Multifuge X3/X3R X3 F/X3 F R Centrifuge Series.
Frequently Bought Together