ContactUsLinkComponent
Fiberlite™ F15-6×100y Fixed-Angle Rotor, Thermo Scientific
Fiberlite™ F15-6×100y Fixed-Angle Rotor, Thermo Scientific
Certificates
undefined
Fiberlite™ F15-6×100y Fixed-Angle Rotor, Thermo Scientific
Specifications
  • Catalog No:
  • CA97040-312
  • Rotor type:
  • Fixed-Angle
  • Angle:
  • 25°
  • Ext.Ø×L:
  • 38 x 105 mm