Rhodium ICP Standard, 1 ml = 1 mg Rh (1,000 ppm Rh)RhCl₃ in 3% HCl, Ricca Chemical Company
RCPRH1KH-500
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:3% HCl
- Size:500 ml
- Cat. no.:RCPRH1KH-500
- Element:Rhodium
- Supplier No.:RPRH1KH0-500N