Rhenium ICP Standard, 1 ml = 1 mg Re (1000 ppm Re) NH₄ ReO₄ in Water, RICCA Chemical Company
RCPRE1KW-500
Small Business Enterprise
:
- Concentration:1000 µg/ml
- Matrix:H₂O
- Size:500 ml
- Cat. no.:RCPRE1KW-500
- Element:Rhenium
- Supplier No.:RPRE1KW0-500N