Mercury ICP Standard, 1 mL = 1 mg Hg (1,000 ppm Hg)Hg in 3% HNO3, Ricca Chemical Company
Catalog # RCPHG1KN-500
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:3% HNO₃
- Size:500 ml
- Cat. no.:RCPHG1KN-500
- Element:Mercury
- Supplier No.:RPHG1KN0-500N