Order Entry
Canada
ContactUsLinkComponent
 
Scandium ICP Standard, 1 ml = 0.01 mg Sc (10 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSSC10N-100
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Scandium ICP Standard, 1 ml = 0.01 mg Sc (10 ppm Sc)Sc₂O₃ in 3% HNO₃, Ricca Chemical Company
Catalog # RCMSSC10N-100
Supplier:  Ricca Chemical
Supplier Number:  RMSSC10N-100N
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml
  • Matrix:
    3% HNO₃
  • Size:
    100 ml
  • Cat. no.:
    RCMSSC10N-100
  • Element:
    Scandium
  • Supplier No.:
    RMSSC10N-100N

Specifications

About this item