Eriochrome Black T 1% (w/v) in 20% (v/v) triethanolamine / 80% (v/v) isopropyl alcohol for hardness of water determinations
RC9301-4
Small Business Enterprise
:
- Size:120 ml
- Cat. no.:RC9301-4
- Supplier No.:R9301000-120C
- Appearance:Dark blue, alcohol odor
- Suitability for Use:Wine Red (excess Ca or Mg) - Pure Blue (excess EDTA)
:
:9,III
:461.39 g/mol
:3077
:1787-61-7
:C₂₀H₁₂N₃NaO₇S
:MFCD00003935