Sulfate Standard Stock Solution, 1000 ppm SO₄²⁻, Ricca Chemical Company
Catalog # RC8109-4
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:1000 ppm
- Description:Sulfate
- Size:20 L
- Cat. no.:RC8109-4
- Contents:K₂SO₄ in H₂O
- Supplier No.:R8109000-120A